Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLX 647 dihydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent dual KIT/Fms inhibitor (IC50 values are 16 and 28 nM respectively). Inhibits BCR-FMS cell proliferation and osteoclast differentiation in vitro (IC50 values are 92 and 170 nM respectively). Attenuates LPS-induced TNFα and IL-6 release, mast cell activation and macrophage kidney proliferation in vivo.
Specifications
Color | White |
Quantity | 10 mg |
Formula Weight | 455.3 |
Percent Purity | >98% |
Physical Form | Solid |
Chemical Name or Material | PLX 647 dihydrochloride |
Safety and Handling
Recommended Storage | Desiccate at RT |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction