Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLXDC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520467
Description
A suggested positive control is human colon tissue lysate. PA5-20467 can be used with blocking peptide PEP-0584.
May play a role in tumor angiogenesis.
Specifications
PLXDC2 | |
Polyclonal | |
Unconjugated | |
PLXDC2 | |
1200007L24Rik; 5430431D22Rik; AI646728; AU022916; AV231634; plexin domain containing 2; plexin domain-containing protein 2; PLXDC2; TEM7R; tumor endothelial marker 7 related; tumor endothelial marker 7-related protein; UNQ2514/PRO6003 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
361282, 67448, 84898 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q6UX71, Q9DC11 | |
PLXDC2 | |
A 17 amino acid peptide from near the carboxy terminus of human Plxdc2. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction