Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLXNC1 Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547730
Description
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse Plexin A1, A2, A3, recombinant human Plexin B1, B2, and D1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Receptor for SEMA7A, for smallpox semaphorin A39R, vaccinia virus semaphorin A39R and for herpesvirus Sema protein. Binding of semaphorins triggers cellular responses leading to the rearrangement of the cytoskeleton and to secretion of IL6 and IL8. {ECO:0000250, ECO:0000269; PubMed:20727575}.
Specifications
PLXNC1 | |
Polyclonal | |
Unconjugated | |
PLXNC1 | |
2510048K12Rik; AW742158; CD232; CD232 antigen; plexin (semaphorin receptor); plexin C1; plexin-C1; PLXNC1; PLXN-C1; receptor for viral semaphorin protein; receptor for virally-encoded semaphorin; Vespr; VESPRplexin-C1; viral-encoded semaphorin protein receptor; virus-encoded semaphorin protein receptor | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
54712 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9QZC2 | |
PLXNC1 | |
CHO-derived recombinant mouse Plexin C1 Ala35-Thr950. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction