Learn More
Epredia™ PrintMate™ Embedding Cassettes
PrintMate Embedding Cassettes are designed for superior ease of use and print clarity.  For use with routine specimens and biopsies wrapped in tissue or sponges.
Manufacturer:  Thermo Scientific™ A84210049
 View more versions of this product
Catalog No. A84210049
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Available in tubes or loose packed, PrintMate Embedding Cassettes protect routine histology specimens.
Designed for performance and user convenience, PrintMate Embedding Cassettes have been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems.  The even, textured printing face provides the optimal surface for high-quality printing every time.  For additional information, the right and left sides are textured for clear handwriting with suitable pens.  The lid catches have been adjusted for ease of use with low opening and closing force, while still giving the security needed while processing.
Available in tubes for automatic printing or loose for manual use, and in twelve colors for enhanced workflow identification.
PrintMate Embedding Cassettes are also available without lids, for economical use with reuseable metal lids.
Specifications
Specifications
Embedding Biopsy Cassettes, Round Pore
Yellow
1000
Yes