Learn More
PM20D2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 177051AP
Catalog No. 50-173-1731
Add to cart
Description
Description
PM20D2 belongs to the peptidase M20A family. The function of PM20D2 remains unknown.
Specifications
Specifications
PM20D2
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A3KG59, Q8IYS1
ACY1L2, Aminoacylase 1 like protein 2, PM20D2
Rabbit
IgG
150 μL
-20°C
Primary
135293, 242377, 313130
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PM20D2
Liquid
Pm20d2
PM20D2 Fusion Protein Ag11991
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat