Learn More
PMEL Mouse anti-Human, Clone: OTI6D5, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF803133
Catalog No. 50-166-6341
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Melanoma gp100 a melanosomal matrix protein whose expression is closely correlated with cellular melanin content and which is a frequent melanoma tumor antigen recognized by cytotoxic T lymphocytes. It is a likely candidate gene for some cases of human oculocutaneous albinism not associated with known albinism loci.
Specifications
Specifications
PMEL
Monoclonal
Unconjugated
PMEL
Lyophilized
PMEL
Human recombit protein fragment corresponding to amino acids 25-467 of human SILV produced in SF9 cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI6D5
PBS with 8% trehalose and no preservative; pH 7.3
P40967
D12S53E, ME20, ME20-M, ME20M, P1, P100, PMEL17, SI, SIL, SILV, gp100
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
6490