Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMEL17/SILV Mouse anti-Human, Equine, PE/Cy5.5, Clone: NKI-beteb, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP233172PECY55

Catalog No. N233172PEC5

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PMEL17/SILV Monoclonal antibody specifically detects PMEL17/SILV in Human, Equine samples. It is validated for Flow Cytometry.
Specifications

Specifications

PMEL17/SILV
NKI-beteb
Flow Cytometry
Conjugated, Purified
PMEL
Lymph node pigmented melanoma metastases extract was used as the immunogen for the gp100 NKI-beteb antibody
Protein A purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
By immunohistochemistry, it specifically recognizes a protein in melanocytes and melanomas. This MAb reacts with junctional and blue nevus cells and variably with fetal and neonatal melanocytes. Intradermal nevi, normal adult melanocytes, and non-melanocytic cells are negative. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin.This Mab labels formalin-fixed, paraffin-embedded melanomas and other tumors showing melanocytic differentiation.
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
D12S53EP1, gp100, ME20, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17P100, premelanosome proteinME20M, SI, SIL, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17
Mouse
IgG2b
0.1 mL
Primary
6490.0
Equine, Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit