Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PMFBP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569495
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
May play a role in sperm morphology especially the sperm tail and consequently affect fertility. May also be involved in the general organization of cellular cytoskeleton.
Specifications
PMFBP1 | |
Polyclonal | |
Unconjugated | |
PMFBP1 | |
1700016D22Rik; F77; LOW QUALITY PROTEIN: polyamine-modulated factor 1-binding protein 1; ODF3; outer dense fiber ODF3; outer dense fiber protein 3; PMF-1 binding protein; PMF-1-binding protein; PMFBP1; polyamine modulated factor 1 binding protein 1; polyamine-modulated factor 1-binding protein 1; sperm tail associated protein; sperm tail-associated protein; Stap | |
Rabbit | |
Affinity chromatography | |
RUO | |
83449 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TBY8 | |
PMFBP1 | |
synthetic peptide directed towards the middle region of human PMFBP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction