Learn More
PMI™ Max Wear Static Rope
For working under extreme environments and rugged terrain
Manufacturer:  Pigeon MountainSupplier Diversity Partner RR125OW061M
 View more versions of this product
Catalog No. 19-817-097
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Orange/White
1.25cm
Working under extreme environments and rugged terrain
61m
44
Static rope
0.5 in.
2.1% at 300 lbf., 3.4% at 600 lbf., 5.8% at 1000 lbf., elongation at 10% MBS: 5.6%
2400 in.
9890
0.22 lb.