Learn More
PMI™ Pulleys and Carabiners
Manufacturer:  Pigeon MountainSupplier Diversity Partner SM21001
 View more versions of this product
Catalog No. 19-817-157
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
5.04 x 2.97 in. (12.8 x 7.5cm)
Zinc plated Steel
10341 lbf.
Stainless Steel gates and locking sleeves
5.04 in.
2.97 in.
1.20 in.
10.6 oz.
Carabiner
Large
Screw lock
NFPA 1983, G
12.8cm
7.5cm
3.05cm
300g