Learn More
PMI™ RA/SMC Stainless-Steel Pulleys
Manufacturer:  Pigeon MountainSupplier Diversity Partner SM150000
 View more versions of this product
Catalog No. 19-816-991
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
4.6 x 2.5 x 1.4 in. (11.6 x 6.4 x 3.6cm)
Stainless Steel
NFPA 1983, T
4.6 in.
2.5 in.
1.4 in.
8.6 oz.
Single Pulley
6519 lbf.
13mm dia. Rope
11.6cm
6.4cm
3.6cm
244g