Learn More
PMI™ Tubular Webbings
For commercial versions of military specifications
Manufacturer:  Pigeon MountainSupplier Diversity Partner WB59021
 View more versions of this product
Catalog No. 19-817-172
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Dark green
Commercial versions of military specifications. For all rigging uses: slings, anchorages, even hasty seats.
92m
4000 lbf
100 yard spool
2.5cm
300 ft. x 1 in. (92m x 2.5cm)
300 ft.
Nylon
18 kN
1 in.
Tubular Webbing