Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PML-5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5120613

This item is not returnable.
View return policy
Description
Positive test controls include: Rat spleen, Rat lung, Rat kidney. The target is usually found in the following locations: Cytoplasm, Cytoplasmic side, Early endosome membrane, Endoplasmic reticulum membrane, Nucleus, PML body, Peripheral membrane protein, nucleolus, nucleoplasm.
PML-5 Polyclonal antibody specifically detects PML-5 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
PML-5 | |
Polyclonal | |
Unconjugated | |
PML | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18854, 315713 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
1.03 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q60953 | |
A synthetic peptide of human PML. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title