Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ PML Protein Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals™ NBP330289
Description
PML Protein Polyclonal antibody specifically detects PML Protein in Human samples. It is validated for Simple Western,Immunohistochemistry,Immunoprecipitation,Western BlotSpecifications
PML Protein | |
Polyclonal | |
Tris-buffered Saline 0.1% BSA | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Simple Western, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
MYLPP8675, promyelocytic leukemia, Promyelocytic leukemia protein, RING finger protein 71, RNF71probable transcription factor PML, TRIM19promyelocytic leukemia, inducer of, tripartite motif protein TRIM19, Tripartite motif-containing protein 19 | |
between 375 and 425 | |
20 μg | |
Cancer, Zinc Finger | |
5371 | |
Store at 4°C. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction