Learn More
BrandTech™ VITLAB™ PMP Wide Mouth Erlenmeyer Flasks
VITLAB wide-mouth Erlenmyer flasks have screw caps for sample integrity
Manufacturer:  BrandTech™ V66795
 View more versions of this product
Catalog No. 14-379-192
Add to cart
Description
Description
  • Neck also accomodates NS stoppers
Specifications
Specifications
Autoclavable
3.38 oz.
Transparent
100 mL
20 mL
GL40
PMP
Printed scale
PMP
34/35