Learn More
BrandTech™ VITLAB™ PMP Wide Mouth Erlenmeyer Flasks
VITLAB wide-mouth Erlenmyer flasks have screw caps for sample integrity
Manufacturer:  BrandTech™ V66895
 View more versions of this product
Catalog No. 14-379-194
Add to cart
Description
Description
  • Neck also accomodates NS stoppers
Specifications
Specifications
Autoclavable
8.45 oz.
Transparent
250 mL
50 mL
GL52
PMP
Printed scale
PMP
45/40