Learn More
Aviva Systems Biology Corp PMS2 Antibody - middle region (ARP56117_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56117_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PMS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PMS2. Alternate names: HNPCC4, PMS2CL, PMSL2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat, Yeast
Catalog No. 50-014-61725