Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMS2 Mouse anti-Human, Clone: 163C1251, Abnova™

Mouse monoclonal antibody raised against synthetic peptide of human PMS2.

Manufacturer:  Abnova Corporation MAB12955

Catalog No. 89-934-785


Add to cart

Description

Description

This gene is one of the PMS2 gene family members found in clusters on chromosome 7. The product of this gene is involved in DNA mismatch repair. It forms a heterodimer with MLH1 and this complex interacts with other complexes bound to mismatched bases. Mutations in this gene are associated with hereditary nonpolyposis colorectal cancer, Turcot syndrome, and are a cause of supratentorial primitive neuroectodermal tumors. Alternatively spliced transcript variants have been observed for this gene. [provided by RefSeq]

Sequence: SSLAKRIKQLHHEAQQS
Specifications

Specifications

PMS2
163C1251
Mouse monoclonal antibody raised against synthetic peptide of human PMS2.
In PBS (0.02% sodium azide).
HNPCC4/PMS2CL/PMSL2
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
PMS2
A synthetic peptide corresponding to amino acids 623-639 of human PMS2.
Protein G purification
RUO
Primary
5395
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit