Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMS2 Rabbit anti-Human, Clone: SY08-09, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267069
Description
PMS2 Monoclonal antibody specifically detects PMS2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
PMS2 | |
SY08-09 | |
Unconjugated | |
Purified | |
DNA mismatch repair protein PMS2, EC 3.1, H_DJ0042M02.9, HNPCC4postmeiotic segregation increased (S. cerevisiae) 2, mismatch repair endonuclease PMS2, PMS1 protein homolog 2, PMS2 postmeiotic segregation increased 2 (S. cerevisiae), PMSL2PMS2CL | |
PMS2 | |
Full length protein | |
Monoclonal | |
Protein A purified | |
Cancer, Cell Cycle and Replication, DNA Repair, Mismatch Repair | |
Human |
Western Blot,Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunoprecipitation | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
5395 | |
Rabbit | |
IgG | |
Primary | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title