Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMS2L5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549101

Catalog No. PA549101


Add to cart

Specifications

Specifications

PMS2L5
Polyclonal
PBS with 0.09% sodium azide
A8MQ11
Postmeiotic segregation increased 2-like protein 5, Postmeiotic segregation increased protein 4, Postmeiotic segregation increased protein 7, Putative postmeiotic segregation increased 2 pseudogene 5, PMS2P5, PMS2L5, PMS4, PMS7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5383
Western Blot
Unconjugated
PMS2P5
Liquid
PMS2P5
KLH conjugated synthetic peptide between 82-107 amino acids from the C-terminal region of human PMS2L5.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit