Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMVK/phosphomevalonate kinase Mouse anti-Human, Rat, Biotin, Clone: OTI2F9, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273484B
Description
PMVK/phosphomevalonate kinase Monoclonal specifically detects PMVK/phosphomevalonate kinase in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
PMVK/phosphomevalonate kinase | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
PMVK | |
Full length recombinant protein of human PMVK/phosphomevalonate kinase (NP_006547) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG2a |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI2F9 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
hPMK, HUMPMKI, phosphomevalonate kinase, PMKA, PMKase, PMKEC 2.7.4.2, PMKI | |
Mouse | |
Affinity Purified | |
Cancer, metabolism | |
10654.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title