Tocris™ PMX 464

Putative inhibitor of the Trx-TrxR system; antiproliferative

Manufacturer: Tocris™ 4996/50

 View more versions of this product

Catalog No. 49-965-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors PMX 464
    Purity >98
    Molecular Weight 243.28
    Quantity 50mg
    Product Type PMX 464

    Putative inhibitor of the thioredoxin-thioredoxin reductase (Trx-TrxR) system. Shown to inhibit Trx and induce a G1/S block in HT29 cells; inhibits cell proliferation in various colorectal cancer cell lines and MCF7 cells. Also elicits an anti-inflammatory response in A549 cells.

    • Chemical Name: 4-(2-Benzothiazolyl)-4-hydroxy-2,5-cyclohexadien-1-one
    • Formula: C13H9NO2S
    • CAS Number: 485842-97-5
    • Solubility: Soluble to 100mM in DMSO and to 100mM in ethanol
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit