Learn More
Tocris Bioscience™ PMX 464
Putative inhibitor of the Trx-TrxR system; antiproliferative
Supplier: Tocris Bioscience™ 4996/10
This item is not returnable.
View return policy
Description
PMX 464 is a putative inhibitor of the thioredoxin-thioredoxin reductase (Trx-TrxR) system. Shown to inhibit Trx and induce a G1/S block in HT29 cells; inhibits cell proliferation in various colorectal cancer cell lines and MCF7 cells. Also elicits an anti-inflammatory response in A549 cells.Specifications
>98% | |
PMX 464 | |
PMX 464 |
10 mg | |
243.28 |