Learn More
Pyramex™ PMXTREME™ LED Safety Eyewear
Clear lens with black frame and pivoting LED temples
$114.10 - $123.00
Specifications
Certifications/Compliance ANSI Z87.1
Weight (Metric) 50 g
Frame Color Black
Frame Material Nylon
Lens Material Polycarbonate
View More Specs
Products 2
Catalog Number Mfr. No. Lens Type Price Quantity & Availability  
Catalog Number Mfr. No. Lens Type Price Quantity & Availability  
19-155-778 View Documents Pyramex™
SB6310SPLED
Anti-UV Pack of 6 for $114.10
Add to cart
19-155-783 View Documents Pyramex™
SB6310STPLED
Anti-UV Pack of 6 for $123.00
Add to cart
Description
Description
  • Built-in rubber nose piece
  • Rubber temple tips keep eyewear secure for the active worker
  • The L1154 alkaline batteries provides up to 50 hours of use before replacing
  • Easy to use push button on/off switches are located at the top of each temple
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • The super bright pivoting LED bulbs (3 lumens per side) help you to see in dark places while keeping your hands free to do other task
  • Three batteries per temples (six total) are included
  • Specifications
    Specifications
    ANSI Z87.1
    Black
    Polycarbonate
    50 g
    Nylon
    Clear