Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PNAS4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542033
Description
Peptide sequence: SCLPKEWLTP AALQSSVSQE LQDELEEAED AAASASVAST AAGSRPGRHT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
PNAS4 is a highly conserved protein that shares high homology from plants to animals. It was initially identified as a putative apoptosis-related protein in the human acute promyelocytic leukemia cell line NB4. PNAS4 is activated during the early response to DNA damage and can inhibit proliferation via apoptosis when overexpressed in some tumor cells such as U2OS, SKOV3, and A549. PNAS4 inhibits tumor cell proliferation through the following mechanisms: (i) overexpression of PNAS4 causes S phase arrest by regulating the expression of cell cycle-related proteins and (ii) PNAS4 induces apoptosis through the mitochondrial apoptosis pathway. Recent evidence has shown that PNAS4 may be involved in the genesis of some cancers and could be a potential candidate for lung cancer therapy alone or in combination with gemcitabine. At least two isoforms of PNAS4 are known to exist.
Specifications
PNAS4 | |
Polyclonal | |
Unconjugated | |
DESI2 | |
5830417C01Rik; C1orf121; cb238; CGI-146; DESI; DESI1; desi2; DeSI-2; desumoylating isopeptidase 1; desumoylating isopeptidase 2; Deubiquitinase DESI2; Fam152a; family with sequence similarity 152, member A; fi12d12; hypothetical protein LOC445138; pnas4; PNAS-4; PPPDE peptidase domain containing 1; PPPDE peptidase domain-containing protein 1; Pppde1; Protein FAM152A; sb:cb238; si:ch211-132e22.3; wu:fi12d12; zgc:100860 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51029 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BSY9 | |
DESI2 | |
Synthetic peptide directed towards the C-terminal of human DESI2 (aa 143-192). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction