Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pan Endothelial Cell Marker Rat anti-Mouse, Alexa Fluor 594, Clone: RM0061-4J13, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP212438AF594
Description
Pan Endothelial Cell Marker Monoclonal antibody specifically detects Pan Endothelial Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Pan Endothelial Cell Marker | |
RM0061-4J13 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
Cell lysates of mouse endothelial cells | |
Protein G purified | |
Store at 4°C in the dark. | |
Monoclonal |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor™ 594 | |
50 mM sodium borate with 0.05% sodium azide | |
Rat | |
IgG2 | |
0.1 mL | |
Primary | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title