Learn More
Fisher Science Education™ Pneumatic Fabric ESD Chairs with Adjustable Arms and Foot Ring, High Bench Height Products for Science Education
Fabric ESD chairs feature proper grounding, conductive, and dissipative static control materials. Fisher Science Education Pneumatic Fabric ESD Chairs with Adjustable Arms and Foot Ring, High Bench Height have been tested to meet ANSI/ESD-STM-12.1-2006.
$559.00 - $590.00
Specifications
Base Style 5-leg polished cast aluminum
Adjustable (Height) 26 to 35.5 in. (66.04 to 90.17cm)
Material Fabric (Static-dissipating, Heavy Duty)
Seating Type Pneumatic ESD Fabric Chair
Width (English) Seat 19 in.
View More Specs
Products 8
Catalog Number Color With Tilt Foot Type Price Quantity & Availability  
Catalog Number Color With Tilt Foot Type Price Quantity & Availability  
S36303 Products for Science Education View Documents Black No ESD, non-marring bell glides Each for $559.00
Add to cart
S36304 Products for Science Education View Documents Blue No ESD, non-marring bell glides Each for $559.00
Add to cart
S36306 Products for Science Education View Documents Black Yes ESD, non-marring bell glides Each for $590.00
Add to cart
S38464 Products for Science Education View Documents Black No ESD rolling, non-marring casters Each for $559.00
Add to cart
S38465 Products for Science Education View Documents Blue No ESD rolling, non-marring casters Each for $559.00
Add to cart
S36208 Products for Science Education View Documents Blue Yes ESD, non-marring bell glides Each for $590.00
Add to cart
S36177 Products for Science Education View Documents Black Yes ESD rolling, non-marring casters Each for $590.00
Add to cart
S36178 Products for Science Education View Documents Blue Yes ESD rolling, non-marring casters Each for $590.00
Add to cart
Description
Description
  • 6-way adjustable arm rests fit all possible ergonomic seating configurations. Adjustable arm rests move up and down, in and out, and twist toward or away from the user
  • 20 in. (50.8cm) diameter chrome
  • Optional seat tilt provides 8° tilt
Specifications
Specifications
5-leg polished cast aluminum
Fabric (Static-dissipating, Heavy Duty)
19 in.
26 to 35.5 in. (66.04 to 90.17cm)
Pneumatic ESD Fabric Chair
Yes