Learn More
Fisher Science Education™ Pneumatic Fabric ESD Chairs with Foot Ring, High Bench Height Products for Science Education
Fabric ESD chairs feature proper grounding, conductive, and dissipative static control materials. Fisher Science Education Pneumatic Fabric ESD Chairs with Foot Ring, High Bench Height have been tested to meet ANSI/ESD-STM-12.1-2006.
$469.00 - $500.00
Specifications
Base Style 5-leg polished cast aluminum
Adjustable (Height) 26 to 35.5 in. (66.04 to 90.17cm)
Material Fabric (Static-dissipating, Heavy Duty)
Seating Type Pneumatic ESD Fabric Chair
Width (English) Seat 19 in.
View More Specs
Products 7
Catalog Number Color With Tilt Foot Type Price Quantity & Availability  
Catalog Number Color With Tilt Foot Type Price Quantity & Availability  
S36379 Products for Science Education View Documents Blue Yes ESD, non-marring bell glides Each for $500.00
Add to cart
S36201 Products for Science Education View Documents Black Yes ESD, non-marring bell glides Each for $500.00
Add to cart
S38567 Products for Science Education View Documents Blue Yes ESD rolling, non-marring casters Each for $500.00
Add to cart
S38570 Products for Science Education View Documents Blue No ESD rolling, non-marring casters Each for $469.00
Add to cart
S38571 Products for Science Education View Documents Black Yes ESD rolling, non-marring casters Each for $500.00
Add to cart
S38600 Products for Science Education View Documents Black No ESD rolling, non-marring casters Each for $469.00
Add to cart
S36183 Products for Science Education View Documents Black No ESD, non-marring bell glides Each for $469.00
Add to cart
Description
Description
  • 20 in. (50.8cm) diameter chrome
  • Optional seat tilt provides 8° forward and 3° backward locking tilt for ergonomic posture support
  • Contoured waterfall cushion and an adjustable back rest with lumbar support for increased com
Specifications
Specifications
5-leg polished cast aluminum
Fabric (Static-dissipating, Heavy Duty)
19 in.
26 to 35.5 in. (66.04 to 90.17cm)
Pneumatic ESD Fabric Chair
Yes