Learn More
Fisher Science Education™ Pneumatic Polyurethane Chair with Medium Back and Adjustable Arms, High-Bench Height Products for Science Education
14.5 in. high back provides more back support. Fisher Science Education Pneumatic Polyurethane Chair with Medium Back and Adjustable Arms, High-Bench Height provide proper seat high range for high work surfaces.
$364.00 - $501.00
Specifications
Color Black
Adjustable (Height) 24.5 to 33.5 in. (62.23 to 85.09cm)
Material Polyurethane
Width (English) Seat 16 in.
Width (English) Back 15.5 in.
View More Specs
Products 10
Catalog Number Wheel Type With Tilt Base Style Price Quantity & Availability  
Catalog Number Wheel Type With Tilt Base Style Price Quantity & Availability  
S36198 Products for Science Education View Documents Black, non-marring bell glides Yes 5-leg nylon reinforced fiberglass Each for $396.00
Add to cart
S36406 Products for Science Education View Documents Black, non-marring bell glides No 5-leg polished cast aluminum Each for $455.00
Add to cart
S36407 Products for Science Education View Documents Black, non-marring bell glides Yes 5-leg polished cast aluminum Each for $487.00
Add to cart
S36700 Products for Science Education View Documents Chrome hooded, rolling, non-marring casters Yes 5-leg black powder coated steel base with fixed foot ring Each for $424.00
Add to cart
S36809 Products for Science Education View Documents Chrome hooded, rolling, non-marring casters No 5-leg polished cast aluminum Each for $469.00
Add to cart
S38460 Products for Science Education View Documents Black, non-marring bell glides No 5-leg nylon reinforced fiberglass Each for $364.00
Add to cart
S37047 Products for Science Education View Documents Chrome hooded, rolling, non-marring casters No 5-leg nylon reinforced fiberglass Each for $378.00
Add to cart
S38447 Products for Science Education View Documents Black, non-marring bell glides Yes 5-leg black powder coated steel base with fixed foot ring Each for $410.00
Add to cart
S38505 Products for Science Education View Documents Chrome hooded, rolling, non-marring casters Yes 5-leg nylon reinforced fiberglass Each for $410.00
Add to cart
S38701 Products for Science Education View Documents Chrome hooded, rolling, non-marring casters Yes 5-leg polished cast aluminum Each for $501.00
Add to cart
Description
Description
  • Polyurethane material is self-skinning, chemical- and puncture-resistant. It is easily cleaned and soil resistant making it ideal for almost any environment
  • 6-way adjustable arm rests support a variety of arm placements for proper ergonomics. Arm rests move up and down, in and out, and twist toward or away from the user
  • Choose from chrome hooded rolling, non-marring casters (suitable for both hard and carpeted floors) or black, non-marring bell glides (suitable for both hard and carpeted floors) on a polished cast aluminum base
  • Optional seat tilt provides 8° forward and 3° backward locking tilt for ergonomic posture support
Specifications
Specifications
Black
Polyurethane
15.5 in.
24.5 to 33.5 in. (62.23 to 85.09cm)
16 in.
14.5 in.