Learn More
Fisher Science Education™ Pneumatic Polyurethane Clean Room Chairs, Desk Height
Clean room chairs have been tested to Fed Std.209E and ISO 14644-1 requirements (Class10/ISO Class 4).
$316.00 - $360.00
Products 6
Catalog Number Back Style Adjustable Wheel Type Price Quantity & Availability  
Catalog Number Back Style Adjustable Wheel Type Price Quantity & Availability  
S36245 Products for Science Education View Documents Standard Height Chrome Casters Each for $316.00
Add to cart
S36362 Products for Science Education View Documents Each for $360.00
Add to cart
S36606 Products for Science Education View Documents Each for $328.00
Add to cart
S38461 Products for Science Education View Documents Standard Seat Height, Tilt Seat Chrome Casters Each for $347.00
Add to cart
S38654 Products for Science Education View Documents Each for $328.00
Add to cart
S38655 Products for Science Education View Documents Each for $360.00
Add to cart
Description
Description
Polyurethane chairs are fully washable, making them ideal for almost any environment.
Certifications
Tested to Fed Std.209E and ISO 14644-1 requirements (Class10/ISO Class 4).