Learn More
Fisher Science Education™ Pneumatic Polyurethane Clean Room Chairs, Medium Bench Height
Clean room chairs have been tested to Fed Std.209E and ISO 14644-1 requirements (Class10/ISO Class 4).
$342.00 - $405.00
Products 12
Catalog Number Back Style Adjustable Wheel Type Price Quantity & Availability  
Catalog Number Back Style Adjustable Wheel Type Price Quantity & Availability  
S36207 Products for Science Education View Documents Each for $372.00
Add to cart
S36240 Products for Science Education View Documents Each for $373.00
Add to cart
S36368 Products for Science Education View Documents Each for $342.00
Add to cart
S36450 Products for Science Education View Documents Each for $405.00
Add to cart
S36464 Products for Science Education View Documents Each for $342.00
Add to cart
S36596 Products for Science Education View Documents Standard Height, Seat Tilt, Foot Ring Bell Glides Each for $377.00
Add to cart
S36822 Products for Science Education View Documents Standard Height, Seat Tilt, Foot Ring Chrome Casters Each for $392.00
Add to cart
S36944 Products for Science Education View Documents Each for $373.00
Add to cart
S37004 Products for Science Education View Documents Standard Height, Foot Ring Chrome Casters Each for $361.00
Add to cart
S37048 Products for Science Education View Documents Each for $405.00
Add to cart
S38451 Products for Science Education View Documents Standard Height, Foot Ring Bell Glides Each for $347.00
Add to cart
S38698 Products for Science Education View Documents Each for $372.00
Add to cart
Description
Description
Polyurethane chairs are fully washable, making them ideal for almost any environment.
Certifications
Tested to Fed Std.209E and ISO 14644-1 requirements (Class10/ISO Class 4).