Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNLIP Mouse anti-Human, Mouse, Clone: GT1777, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527849

Catalog No. PIMA527849


Add to cart

Description

Description

Positive Control: mouse pancreas Predicted Reactivity: Human (100%), Pig (90%), Rabbit (84%), Bovine (85%), Guinea pig (82%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

PNLIP encodes a member of the lipase family of proteins. The encoded enzyme is secreted by the pancreas and hydrolyzes triglycerides in the small intestine, and is essential for the efficient digestion of dietary fats. Inhibition of the encoded enzyme may prevent high-fat diet-induced obesity in mice and result in weight loss in human patients with obesity. Mutations in this gene cause congenital pancreatic lipase deficiency, a rare disorder characterized by steatorrhea.
Specifications

Specifications

PNLIP
Monoclonal
1 mg/ml
PBS with no preservative
P16233, Q6P8U6
PNLIPD, PTL, PNLIP, PL, pancreatic lipase
Mouse
IgG1
100 μl
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5406, 69060
Western Blot
GT1777
Unconjugated
PNLIP
Liquid
Pnlip
Recombit protein encompassing a sequence within the center region of human Pancreatic Lipase. The exact sequence is proprietary.
Protein G
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit