Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNMA5 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548922
Description
This gene encodes a member of the paraneoplastic Ma antigen protein family. These proteins have been implicated in the development of paraneoplastic disorders resulting from an immune response directed against them. Paraneoplastic disorders are the result of an abnormal immune response to a tumor. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq, Oct 2011]
Specifications
PNMA5 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q96PV4 | |
PNMA5 | |
KLH conjugated synthetic peptide between 387-416 amino acids from the C-terminal region of human PNMA5. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PNMA5 | |
Paraneoplastic antigen-like protein 5, Tumor antigen BJ-HCC-25, PNMA5, KIAA1934 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114824 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction