Learn More
Aviva Systems Biology Corp PNPO Antibody - N-terminal region (ARP58515_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58515_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PNPO. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PNPO. Alternate names: FLJ10535, PDXPOApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62336