Learn More
PNPT1 Mouse anti-Human, DyLight 350, Clone: 3H5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243712UV
Catalog No. NBP243712UV
Add to cart
Description
Description
PNPT1 Monoclonal antibody specifically detects PNPT1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
PNPT1
3H5
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
PNPT1
The immunogen used to generate this antibody corresponds to human PNPase.
100 ul
Primary
87178
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
DKFZp762K1914, EC 2.7.7, EC 2.7.7.8, mitochondrial, old-35, OLD35, PNPase, PNPase 1, PNPase old-35, Polynucleotide phosphorylase 1, Polynucleotide phosphorylase-like protein, polyribonucleotide nucleotidyltransferase 1
Mouse
Protein G purified
Store at 4C in the dark.
Monoclonal
Human