Learn More
PNPT1 Mouse anti-Human, HRP, Clone: 3H5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243712H
Catalog No. NBP243712H
Add to cart
Description
Description
PNPT1 Monoclonal antibody specifically detects PNPT1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
PNPT1
3H5
Western Blot
Purified
PNPT1
The immunogen used to generate this antibody corresponds to human PNPase.
100 ul
Primary
87178
Western Blot
HRP
PBS with No Preservative
DKFZp762K1914, EC 2.7.7, EC 2.7.7.8, mitochondrial, old-35, OLD35, PNPase, PNPase 1, PNPase old-35, Polynucleotide phosphorylase 1, Polynucleotide phosphorylase-like protein, polyribonucleotide nucleotidyltransferase 1
Mouse
Protein G purified
Store at 4C in the dark.
Monoclonal
Human