Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNU 22394 hydrochloride, Tocris Bioscience™

Catalog Number 220150
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
15923-42-9
C13H17ClN2
236.743
SAIGGEUWSYESTR-UHFFFAOYSA-N
pnu 22394 hydrochloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole hydrochloride, 6-methyl-1h,2h,3h,4h,5h-azepino 4,5-b indole hydrochloride, 6-methyl-1,2,3,4,5,6-hexahydroazepino 4,5-b indol-6-ium chloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indol, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole-hydrochloride
16759162
6-methyl-2,3,4,5-tetrahydro-1H-azepino[4,5-b]indole;hydrochloride
CN1C2=C(CCNCC2)C3=CC=CC=C31.Cl
This item is not returnable. View return policy
15923-42-9
C13H17ClN2
236.743
SAIGGEUWSYESTR-UHFFFAOYSA-N
pnu 22394 hydrochloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole hydrochloride, 6-methyl-1h,2h,3h,4h,5h-azepino 4,5-b indole hydrochloride, 6-methyl-1,2,3,4,5,6-hexahydroazepino 4,5-b indol-6-ium chloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indol, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole-hydrochloride
16759162
6-methyl-2,3,4,5-tetrahydro-1H-azepino[4,5-b]indole;hydrochloride
CN1C2=C(CCNCC2)C3=CC=CC=C31.Cl

5-HT2C agonist and 5-HT2A/2B partial agonist.

Potent 5-HT2C agonist and partial 5-HT2A/5-HT2B agonist. Non-selective between 5-HT2 receptor subtypes (Ki values are 18, 18 and 66 nM for human recombinant 5-HT2C, 5-HT2A and 5-HT2B receptors respectively). Reduces food intake in rats following subcutaneous administration and displays anorexigenic effects in humans.
Quantity 50 mg
Formula Weight 236.74
Percent Purity >99%
Chemical Name or Material PNU 22394 hydrochloride
Recommended Storage Desiccate at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit