Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNU 96415E, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Antipsychotic agent. Displays high affinity for dopamine D4 and serotonergic 5-HT2A receptors and relatively weak affinity at D2 receptors (Ki values are 3.0, 5.8, 134, 181, 199, 240, 411 and > 678 nM for D4, 5-HT2A, 5-HT1A, α1, D2, D3, D1, α2 and muscarinic receptors respectively). Inhibits exploratory locomotor activity and antagonizes d-amphetamine-induced locomotor stimulation in rats.
Specifications
Quantity | 10 mg |
Formula Weight | 413.36 |
Percent Purity | >98% |
Chemical Name or Material | PNU 96415E |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature