Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNU 96415E, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Antipsychotic agent. Displays high affinity for dopamine D4 and serotonergic 5-HT2A receptors and relatively weak affinity at D2 receptors (Ki values are 3.0, 5.8, 134, 181, 199, 240, 411 and > 678 nM for D4, 5-HT2A, 5-HT1A, α1, D2, D3, D1, α2 and muscarinic receptors respectively). Inhibits exploratory locomotor activity and antagonizes d-amphetamine-induced locomotor stimulation in rats.
Specifications
Quantity | 10 mg |
Formula Weight | 413.36 |
Percent Purity | >98% |
Chemical Name or Material | PNU 96415E |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction