Learn More
Cole-Parmer Kestrel 0820 (GREEN) Pocket Thermo Wind Meter Plus  DFS Item
Manufacturer:  Cole-Parmer 1038211
This convenient handheld meter will measure wind speed and other weather parameters with field selectable units including:
  • Knots (KT)
  • Meters per second (M/S)
  • Kilometers per hour (KM/H)
  • Miles per hour (MPH)
  • Feet per minute (FPM)
  • Beaufort force("B")
Kestrel 2000 - Pocket Wind Meter Features
  • Current Wind Speed
  • Max Wind Gust
  • Average Wind Speed
  • KL008102 - Pocket Thermo Wind Meter Plus Features
  • Wind Speed
  • Temperature
  • Wind Chill
    A number of accessories are available including a portable vane mount and a tripod for longer readings, and a replacement impeller.
Catalog No. 50-631-818