Learn More
Cole-Parmer Kestrel 0820 (GREEN) Pocket Wind Meter, LCD Display  DFS Item
Manufacturer:  Cole-Parmer 1038209
This convenient handheld meter will measure wind speed and other weather parameters with field selectable units including:
  • Knots (KT)
  • Meters per second (M/S)
  • Kilometers per hour (KM/H)
  • Miles per hour (MPH)
  • Feet per minute (FPM)
  • Beaufort force("B")
Kestrel 1000 - Pocket Wind Meter Features
  • Current Wind Speed
  • Max Wind Gust
  • Average Wind Speed
A number of accessories are available including a portable vane mount and a tripod for longer readings, and a replacement impeller.
Catalog No. 50-631-817