Learn More
R3 Safety™ Pocket Zipper Vest Non-distribution product as customer accommodation.
Versatile vest with tough nylon shell and four roomy pockets
Manufacturer:  R3 Safety™ I465ORG04000
 View more versions of this product
Catalog No. 18-999-1254C
Add to cart
This item is not returnable. View return policy
Description
Description
  • With full Velcro™ closures and rustproof Vislon™ zipper
  • Adjustable sides and seven-piece foam design enhance fit and comfort
  • Eyelets for attaching rescue light (not included)
  • SOLAS-grade reflective panels measure 64 sq. in. (413cm2)
  • International orange
Specifications
Specifications
4.0
Flotation Vest
Nylon
Vislon™ zipper
Large
Orange
USCG Type III
Designed with four roomy hook-and-loop pockets to keep gear close, a couple d-rings to hang small tools and eyelets to attach safety equipment