Thermo Scientific™ PocketTip™ D.A.R.T.'S™ (Disposable Automation Research Tips)

Reduce reagent, labware and sample costs while improving data quality through the elimination of intermediate pipetting steps with soft in-tip mixing.

Manufacturer: Thermo Scientific™ 5757250

 View more versions of this product

Catalog No. 14-387-593

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    For Use With (Equipment) Thermo Scientific Matrix Hydra DT
    Product Line D.A.R.T.'S
    Filtered No
    Sterility Sterile
    Packaging 20 racks of 384 tips/case,20 racks of 384 tips/case
    Volume (Metric) 250nL,250nL
    Sterile Yes

    Reduce reagent, labware and sample costs while improving data quality through the elimination of intermediate pipetting steps with soft in-tip mixing. Thermo Scientific™ PocketTip™ D.A.R.T.'s™ achieve lower volume transfer capability due to a combination of the internally molded capillary pocket and the unique surface sealing of the tip.

    Features:

    • Allows existing Thermo Scientific™ Matrix™ automated liquid handlers to accommodate low-volume dispensing for compound screening procedures
    • Requires only minor procedural adjustments to Thermo Scientific™ PlateMate™ and Thermo Scientific™ Hydra™ DT instrumentation
    • Eliminate aqueous dilution steps that can cause precipitation by transferring nanoliter volumes directly to assay plate.
    • Control consistent accuracy and precision during the molding process
    Validation and Compatibility:
    • Purity and straightness validation are performed on compatible instrumentation to ensure only the highest quality.
    • With instrument compatibility range starting from the Matrix Hydra DT, on-the-fly dose response curves or assay development can be set up with simple Hydra DT programming.
    • Higher throughput needs can be met with the Matrix PlateMate 2 x 3 and Catalyst Express or Matrix PlateMate Plus instruments.
    • Sales specialists are available to assist with instrument programming and modifications or instrument validation of any Matrix automated liquid handler.
    Ordering Information:
    • Available in 96- and 384- channel format with 50, 100 and 250nL pocket sizes
    • Sterile format available for sensitive liquid handling procedures

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit