Learn More
Thermo Scientific™ PocketTips™ Pipette Tips
Eliminate time-consuming steps, reduce lab ware usage and accelerate assays with PocketTips™ Pipette Tips.
Specifications
Product Line PocketTip
Filtered Non-filtered
Sterility Non-sterile
View More Specs
Products 15
Catalog Number Mfr. No. Volume (Metric) For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) For Use With (Equipment) Price Quantity & Availability  
14-387-363
View Documents
Thermo Scientific™
FLIPR384B250
250 nL Molecular Devices™ FLIPR 384-Well Head This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-387-364
View Documents
Thermo Scientific™
FLIPR384B50
50 nL Molecular Devices™ FLIPR 384-Well Head This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-387-365
View Documents
Thermo Scientific™
FX384P30100
100 nL Beckman Coulter™ Biomek™ FX This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-387-366
View Documents
Thermo Scientific™
FX384P30250
250 nL Beckman Coulter™ Biomek™ FX This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-387-367
View Documents
Thermo Scientific™
FX384P3050
50 nL Beckman Coulter™ Biomek™ FX This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-387-368
View Documents
Thermo Scientific™
FX96P20100
100 nL Beckman Coulter™ Biomek™ FX This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-387-370
View Documents
Thermo Scientific™
FX96P2050
50 nL Beckman Coulter™ Biomek™ FX This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-387-371
View Documents
Thermo Scientific™
FX96P20500
500 nL Beckman Coulter™ Biomek™ FX This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-387-372
View Documents
Thermo Scientific™
PE384P30100
100 nL Packard™ 384-Well This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-387-373
View Documents
Thermo Scientific™
PE384P3050
50 nL Packard™ 384-Well This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-387-374
View Documents
Thermo Scientific™
PE96P235100
100 nL Packard™ 96-Well This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-387-375
View Documents
Thermo Scientific™
PE96P235250
250 nL Packard™ 96-Well This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-387-376
View Documents
Thermo Scientific™
STAR384P30100
100 nL Hamilton™ MICROLAB™ STAR This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-387-377
View Documents
Thermo Scientific™
STAR384P30250
250 nL Hamilton™ MICROLAB™ STAR This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-387-378
View Documents
Thermo Scientific™
STAR384P3050
50 nL Hamilton™ MICROLAB™ STAR This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Eliminate time-consuming pipetting steps, reduce lab ware usage and accelerate assays such as PCR primer dilutions with user-friendly PocketTips™ Pipette Tips, which are compatible on multiple workstations. The external dimensions of the tip and rack are the same as standard automation tips, requiring minimal modification and set-up. Simple, economical assay miniaturization preserves compounds and reduces reagent costs.
Features:
  • Transfer samples directly to the assay plate
  • Combine sample and assay material with soft in-tip mixing
  • Coefficient of variation (CV) of less than 6% can be achieved for any pocket size
  • Color: Clear
Recommended for:
Cell-based or low DMSO tolerant assays, selectivity assays (P450), potency determinations (IC50), compound storage and delivery from compound libraries, protein and enzyme aqueous based assays, automated clinical diagnostics and kinetic read (FLIPR) assays.
Warranty: A five (5)-year shelf life from the date of manufacture is recommended.
Specifications
Specifications
PocketTip
Non-filtered
Non-sterile
SDS
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.