Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podocalyxin Like, Mouse anti-Human, Clone: PODXL/2185, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP27974920UG

 View more versions of this product

Catalog No. NB010516


Add to cart

Description

Description

Podocalyxin Like Monoclonal specifically detects Podocalyxin Like in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

Podocalyxin Like
PODXL/2185
Unconjugated
10 mM PBS with 0.05% BSA
Purified
PODXL
Recombinant fragment of human Podocalyxin Like(around aa 310-447) (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Podocalyxin Like is a member of the CD34 transmembrane sialomucin family. It is over-expressed on the podocyte foot projections and plays essential roles in kidney development and homeostasis, blood filtration and urine formation. It is also expressed on vascular endothelia, hematopoietic progenitors and a subset of neurons. Overexpression of podocalyxin may be linked to more aggressive tumor behavior. Podocalyxin Like antibody can identify podocytes in the urine (podocyturia) that may indicate glomerular disease, pre-eclampsia, and other kidney pathology.
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
0.2 mg/mL
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
O00592
Gp200MGC138240, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, podocalyxin-like, Podocalyxin-like protein 1
Mouse
IgG1 Kappa
20 ug
Cancer, Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers
Primary
5420
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit