Learn More
AdipoGenSupplier Diversity Partner PODOPHYLLOTOXIN(100MG) DFS Item
Manufacturer:  AdipoGenSupplier Diversity Partner AGCN20049M100
Podophyllotoxin, AdipoGen Life Sciences. Chemical. CAS: 518-28-5. Formula: C22H22O8. MW: 414.4. Isolated from Podophyllum emodi rhizomes. Potent microtubule assembly inhibitor. Anticancer compound. Cell death inducer. DNA topoisomerase II inhibitor. Cell cycle inhibitor at the metaphase. Antiviral and antihelminthic.
Catalog No. 50-114-6239