Learn More
Podoplanin Hamster anti-Mouse, PE, Clone: 8.1.1, BD Pharmingen™
Hamster Monoclonal Antibody
Manufacturer:  BD Biosciences 566390
Catalog No. BDB566390
Add to cart
Description
Description
Podoplanin Monoclonal antibody specifically detects Podoplanin in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Podoplanin
8.1.1
PE
Hamster
0.1 mg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG2
RUO
Primary
Mouse