Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podoplanin Antibody (PDPN/1433), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254347AF350
Description
Podoplanin Monoclonal specifically detects Podoplanin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Podoplanin | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
PDPN | |
Recombinant human Podoplanin protein fragment (around aa 24-126) (exact sequence is proprietary) (Uniprot: Q86YL7) | |
Primary | |
Human | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PDPN/1433 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
Mouse | |
0.1 mL | |
10630 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction