Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podoplanin Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 220991AP

Catalog No. 50-173-3724


Add to cart

Description

Description

Podoplanin (PDPN, T1A, gp38, Aggrus) is a mucin type-1 glycoprotein with a 40-43 kDa molecular weight. Podoplanin is expressed in many tumors and normal cells, especially lymphatic epithelial cells and follicular DCs. Podoplanin localizes in stromal cells of peripheral lymphoid tissue and thymic epithelial cells. As a regulator of the lymphatic endothelium, podoplanin probably plays a role in maintaining the unique shape of podocytes. Podoplanin appears to serve as a ligand for CLEC-2 and expression is positively correlated with tumors expressing greater invasive and metastatic potential. Podoplanin is directly involved in cell migration, aids metastases formation and tumor cell invasion of tissue. Further, it has also been determined that podoplanin expression is induced through Src activation of Cas and, recently, there are indications of podoplanin presence on Th17 cells. Initial studies below indicate the presence of podoplanin on a small but distinct population of CD4+ cells from human PBMC. Dysfunction of the podoplanin protein has been linked to diseases such as lymphangioma and subglottis neoplasm. Podoplanin has also been proposed as a marker for lung injury due to the physiological function of the protein.
Specifications

Specifications

Podoplanin
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q86YL7
AGGRUS, D2 40, D2 40,M2A, D240, Glycoprotein 36, GP36, Gp38, GP40, HT1A 1, M2A, OTS8, PA2.26, PA2.26 antigen, PDPN, Podoplanin, PSEC0025, T1 alpha, T1A, T1A 2
Rabbit
IgG
150 μL
-20°C
Primary
10630
Western Blot
Unconjugated
PDPN
Liquid
PDPN
Podoplanin Fusion Protein Ag17307
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit