Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podoplanin Rat anti-Mouse, Janelia Fluor 549, Clone: pmab-1, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP203955JF549
Description
Podoplanin Monoclonal antibody specifically detects Podoplanin in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
Podoplanin | |
pmab-1 | |
Western Blot, Flow Cytometry | |
Conjugated, Purified | |
PDPN | |
The immunogen is Podoplanin. | |
Protein G purified | |
Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
Primary | |
10630.0 |
Flow Cytometry, Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
Rat | |
IgG2a Kappa | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title